
4-(Isopentyloxy)benzenecarboxylic acid
Catalog # APOSOR12106-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:208.26 g/mol
Melting Pt:142…144 °C
:GHS07
Storage Temperature:Ambient
CAS Number:2910-85-2
Formula:C₁₂H₁₆O₃
MDL Number:MFCD01814579