
Dimethyl-4-iodo-1H-pyrazole-3,5-dicarboxylate 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1027819-68-6
:
Formula:C₇H₇IN₂O₄
MDL Number:MFCD08689740
Melting Pt:176…178 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together