

2-Chloro-4-methylpyrimidine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:13036-57-2
:
Formula:C₅H₅ClN₂
MDL Number:MFCD00054434
:128.56 g/mol
Boiling Pt:93…94 °C/17mmHg
Melting Pt:45…53 °C
Flash Pt:98 °C
:GHS05
:GHS07
Storage Temperature:Freezer
Frequently Bought Together