Order Entry
Czech Republic
ContactUsLinkComponent
 
2,6-Dimethoxyaniline hydrochloride
Catalog # APOSOR11968-1G
Supplier:  Apollo Scientific
CAS Number:  
2,6-Dimethoxyaniline hydrochloride
Catalog # APOSOR11968-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :189.6394 g/mol
Storage Temperature:Ambient
CAS Number:375397-36-7
Formula:C₈H₁₂ClNO₂
MDL Number:MFCD09878313