Order Entry
Czech Republic
ContactUsLinkComponent
2-(3-Bromophenyl)-1-(pyrrolidin-1-yl)ethanone
Irritant
2-(3-Bromophenyl)-1-(pyrrolidin-1-yl)ethanone
Catalog # APOSOR11952-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-(3-Bromophenyl)-1-(pyrrolidin-1-yl)ethanone
Catalog # APOSOR11952-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :GHS07
Storage Temperature:Ambient
CAS Number:951884-73-4
Formula:C₁₂H₁₄BrNO
MDL Number:MFCD09800919