
Methyl-5-bromo-2-chloronicotinate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:78686-79-0
:
Formula:C₇H₅BrClNO₂
MDL Number:MFCD03844848
:250.48 g/mol
Melting Pt:48…52 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together