Order Entry
Czech Republic
ContactUsLinkComponent
4-{2-[4-(4,4,5,5-Tetramethyl[1,3,2]dioxaborolan-2-yl)phenoxy]ethyl}morpholine 96%
Irritant
4-{2-[4-(4,4,5,5-Tetramethyl[1,3,2]dioxaborolan-2-yl)phenoxy]ethyl}morpholine 96%
Catalog # APOSOR11446-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-{2-[4-(4,4,5,5-Tetramethyl[1,3,2]dioxaborolan-2-yl)phenoxy]ethyl}morpholine 96%
Catalog # APOSOR11446-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :333.24 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:690636-28-3
Formula:C₁₈H₂₈BNO₄
MDL Number:MFCD04440857