Order Entry
Czech Republic
ContactUsLinkComponent
Ethyl-8-bromo-5-methylimidazo[1,2-a]pyridine-2-carboxylate 98%
Ethyl-8-bromo-5-methylimidazo[1,2-a]pyridine-2-carboxylate 98%
Catalog # APOSOR11400-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl-8-bromo-5-methylimidazo[1,2-a]pyridine-2-carboxylate 98%
Catalog # APOSOR11400-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :283.1212 g/mol
Storage Temperature:Ambient
CAS Number:135995-45-8
Formula:C₁₁H₁₁BrN₂O₂
MDL Number:MFCD09475853