
4-(2-Methoxyphenoxy)benzenesulphonyl chloride 95%
Catalog # APOSOR11345-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:298.75 g/mol
Melting Pt:54…56 °C
Storage Temperature:Refrigerator
UN:3261
ADR:8,III
:GHS05
CAS Number:690632-30-5
Formula:C₁₃H₁₁ClO₄S
MDL Number:MFCD01631898