

Pyrogallol 98+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:87-66-1
:
Formula:C₆H₆O₃
MDL Number:MFCD00002192
:126.11 g/mol
Boiling Pt:309 °C
Melting Pt:131…135 °C
Density:1.45 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:201-762-9
:GHS07
:GHS08
Frequently Bought Together