Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
1-Ethylpiperidin-3-one hydrochloride 97%
Catalog # APOSOR11245-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-Ethylpiperidin-3-one hydrochloride 97%
Catalog # APOSOR11245-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :163.65 g/mol
Melting Pt:172…173.5 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:41361-28-8
Formula:C₇H₁₄ClNO
MDL Number:MFCD00012793