2',4'-Dihydroxyacetophenone 98%
Catalog # APOSOR1119-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:152.15 g/mol
Melting Pt:143…145 °C
Density:1.18 g/cm³ (25 °C)
Storage Temperature:Ambient
CAS Number:89-84-9
Formula:C₈H₈O₃
MDL Number:MFCD00002279