
4-Bromo-2-nitrotoluene 96%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:60956-26-5
:
Formula:C₇H₆BrNO₂
MDL Number:MFCD00041243
:216.03 g/mol
Boiling Pt:130 °C/12mmHg
Melting Pt:45…48 °C
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:262-536-3
:GHS07
Frequently Bought Together