Bromomethyl methyl ether
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:13057-17-5
- Formula:C₂H₅BrO
- MDL Number:MFCD00000171
- Molecular Weight:124.97 g/mol
- Boiling Pt:27 °C
- Melting Pt:87 °C
- Density:1.531 g/cm³ (25 °C)
- Storage Temperature:Refrigerator
- UN:1992
- ADR:3+6.1,III
6 Options Available Below
Product Details & Documents
- CAS Number:13057-17-5
- Formula:C₂H₅BrO
- MDL Number:MFCD00000171
- Molecular Weight:124.97 g/mol
- Boiling Pt:27 °C
- Melting Pt:87 °C
- Density:1.531 g/cm³ (25 °C)
- Storage Temperature:Refrigerator
- UN:1992
- ADR:3+6.1,III
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



