4-Bromo-1,5-dimethyl-1H-pyrazole-3-carboxylic acid
Catalog # APOSOR111332-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Storage Temperature:Ambient
CAS Number:5775-91-7
Formula:C₆H₇BrN₂O₂
MDL Number:MFCD02090877