Ethyl-2-(6-bromoimidazo[1,2-a]pyridin-2-yl)acetate
Catalog # APOSOR111216-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:283.125 g/mol
Storage Temperature:Ambient
CAS Number:59128-04-0
Formula:C₁₁H₁₁BrN₂O₂
MDL Number:MFCD16172180