1,5-Dimethyl-1H-pyrazole-3-carbohydrazide
Catalog # APOSOR111143-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:154.17 g/mol
Storage Temperature:Ambient
CAS Number:94447-28-6
Formula:C₆H₁₀N₄O
MDL Number:MFCD00221100