
3-(4-Methylpiperazin-1-ylmethyl)benzylamine
Catalog # APOSOR111035-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:GHS07
Storage Temperature:Refrigerator
CAS Number:515162-19-3
Formula:C₁₃H₂₁N₃