6-Nitroimidazo[1,2-a]pyridine hydrochloride
Catalog # APOSOR111033-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Storage Temperature:Ambient
CAS Number:957120-37-5
Formula:C₇H₆ClN₃O₂
MDL Number:MFCD09800971