2-Methyl-6-nitroimidazo[1,2-a]pyridine
Catalog # APOSOR110937-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:177.16 g/mol
Storage Temperature:Ambient
CAS Number:13212-83-4
Formula:C₈H₇N₃O₂
MDL Number:MFCD05863412