
Methyl-4'-bromo-4-nitro[1,1'-biphenyl]-2-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:886361-85-9
:
Formula:C₁₄H₁₀BrNO₄
MDL Number:MFCD08056637
:336.15 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together