
Ethyl-3-bromoimidazo[1,2-a]pyridine-6-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1215504-30-5
:
Formula:C₁₀H₉BrN₂O₂
MDL Number:MFCD12827886
:269.10 g/mol
Melting Pt:95…96 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together