Order Entry
Czech Republic
ContactUsLinkComponent
2-(4-Morpholino)-3-nitropyridine
2-(4-Morpholino)-3-nitropyridine
Catalog # APOSOR110695-5G
Supplier:  Apollo Scientific
CAS Number:  
2-(4-Morpholino)-3-nitropyridine
Catalog # APOSOR110695-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :209.2019 g/mol
Storage Temperature:Ambient
CAS Number:24255-27-4
Formula:C₉H₁₁N₃O₃
MDL Number:MFCD00452814