tert-Butyl-4-[2-(4-formylphenoxy)ethyl]piperazine-1-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1086378-39-3
:
Formula:C₁₈H₂₆N₂O₄
MDL Number:MFCD11042248
:334.42 g/mol
Storage Temperature:Ambient
Frequently Bought Together