Order Entry
Czech Republic
ContactUsLinkComponent
2-Methyl-2H-1,4-benzothiazin-3(4H)-one
2-Methyl-2H-1,4-benzothiazin-3(4H)-one
Catalog # APOSOR110446-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Methyl-2H-1,4-benzothiazin-3(4H)-one
Catalog # APOSOR110446-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :179.237 g/mol
Storage Temperature:Ambient
CAS Number:7028-57-1
Formula:C₉H₉NOS
MDL Number:MFCD02941599