Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(1H-Pyrazol-3-yl)pyridine
4-(1H-Pyrazol-3-yl)pyridine
4-(1H-Pyrazol-3-yl)pyridine
4-(1H-Pyrazol-3-yl)pyridine
APOSOR110289-1G
:
Apollo Scientific
:
4-(1H-Pyrazol-3-yl)pyridine
APOSOR110289-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
145.17 g/mol
:
157…158 °C
:
1.391 g/cm³ (25 °C)
:
GHS07
:
Ambient
:
17784-60-0
:
C₈H₇N₃
:
MFCD00115150