Order Entry
Czech Republic
ContactUsLinkComponent
(2E)-1-(4-Bromophenyl)-3-(dimethylamino)prop-2-en-1-one
(2E)-1-(4-Bromophenyl)-3-(dimethylamino)prop-2-en-1-one
Catalog # APOSOR110236-25G
Supplier:  Apollo Scientific
CAS Number:  
(2E)-1-(4-Bromophenyl)-3-(dimethylamino)prop-2-en-1-one
Catalog # APOSOR110236-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :254.13 g/mol
Melting Pt:80…82 °C
Storage Temperature:Ambient
CAS Number:73387-60-7
Formula:C₁₁H₁₂BrNO
MDL Number:MFCD00121187