
2-Butyl-1H-benzimidazole
Catalog # APOSOR110234-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:174.25 g/mol
Melting Pt:146…148 °C
:GHS07
Storage Temperature:Ambient
CAS Number:5851-44-5
Formula:C₁₁H₁₄N₂
MDL Number:MFCD00714456