
7-Chloroimidazo[1,2-a]pyridine-2-carbohydrazide
Catalog # APOSOR110224-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:210.62 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1215727-61-9
Formula:C₈H₇ClN₄O
MDL Number:MFCD11505066