2-Methylimidazo[1,2-a]pyridine 97%
Catalog # APOSOR110181-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:132.17 g/mol
Storage Temperature:Ambient
CAS Number:934-37-2
Formula:C₈H₈N₂
MDL Number:MFCD02251330