Order Entry
Czech Republic
ContactUsLinkComponent
2-Chloroethyl ethyl sulphide 98%
Flammable
Corrosive
Toxic
Health
2-Chloroethyl ethyl sulphide 98%
Catalog # APOSOR10841-5G
Supplier:  Apollo Scientific
CAS Number:  
Flammable
Corrosive
Toxic
Health
2-Chloroethyl ethyl sulphide 98%
Catalog # APOSOR10841-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
Boiling Pt:156…157 °C
Density:1.07 g/cm³ (25 °C)
Flash Pt:52 °C
Storage Temperature:Refrigerator
EINECS:211-742-1
UN:2929
ADR:6.1+3,II
 :GHS02
 :GHS05
 :GHS06
 :GHS08
CAS Number:693-07-2
Formula:C₄H₉ClS
MDL Number:MFCD00000979