
N-(Bromomethyl)phthalimide 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5332-26-3
:
Formula:C₉H₆BrNO₂
MDL Number:MFCD00005897
:240.06 g/mol
Melting Pt:144…150 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together