
4-Bromo-2,6-dimethylanisole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:14804-38-7
:
Formula:C₉H₁₁BrO
MDL Number:MFCD00143259
:215.09 g/mol
Boiling Pt:117…120 °C/13mmHg
Density:1.347 g/cm³ (25 °C)
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together