
Diethyl dodecylphosphonate
Catalog # APOSOR10707-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:306.42 g/mol
Boiling Pt:168 °C/1.5mmHg
:GHS07
Storage Temperature:Ambient
CAS Number:4844-38-6
Formula:C₁₆H₃₅O₃P
MDL Number:MFCD00015595