
4-tert-Butylphenylhydrazine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:128231-55-0
:
Formula:C₁₀H₁₇ClN₂
MDL Number:MFCD00013387
:200.7084 g/mol
Melting Pt:228 °C (dec.)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together