
Tetrahydropyran-4-methanol 98%
Catalog # APOSOR10585-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:116.16 g/mol
Boiling Pt:105…110 °C/20mmHg
Flash Pt:>61 °C
:GHS07
Storage Temperature:Ambient
CAS Number:14774-37-9
Formula:C₆H₁₂O₂
MDL Number:MFCD00457804