
1-Methylimidazole-4-carboxylic acid methyl ester
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:17289-19-9
:
Formula:C₆H₈N₂O₂
MDL Number:MFCD02179561
:140.1400 g/mol
Melting Pt:101.5…103.5 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together