
1-Methylimidazole-2-carboxylic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:20485-43-2
:
Formula:C₅H₆N₂O₂
MDL Number:MFCD01863421
:126.11 g/mol
Melting Pt:94…98 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together