
Ethyl tetrahydropyran-4-ylacetate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:103260-44-2
:
Formula:C₉H₁₆O₃
MDL Number:MFCD03788560
:172.2215 g/mol
Boiling Pt:73…75 °C/0.2mmHg
:GHS07
Storage Temperature:Ambient
Frequently Bought Together