
5-Bromo-2-chlorobenzoic acid 97+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:21739-92-4
:
Formula:C₇H₄BrClO₂
MDL Number:MFCD00002415
:235.46 g/mol
Melting Pt:157…162 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together