
4-(1H-Pyrazol-4-yl)pyrimidine 95+%
Catalog # APOSOR1031-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:146.15 g/mol
Melting Pt:205…208 °C
:GHS07
Storage Temperature:Ambient
CAS Number:28648-87-5
Formula:C₇H₆N₄
MDL Number:MFCD02579319