
4-(1H-Pyrazol-4-yl)pyridine 95+%
Catalog # APOSOR1030-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:145.1613 g/mol
Melting Pt:195…198 °C
:GHS07
Storage Temperature:Ambient
CAS Number:19959-71-8
Formula:C₈H₇N₃
MDL Number:MFCD00480339