
Tetrabutylammonium bromide 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1643-19-2
:
Formula:C₁₆H₃₆BrN
MDL Number:MFCD00011633
:322.37 g/mol
Melting Pt:101…106 °C
Density:1.15 g/cm³ (25 °C)
EINECS:216-699-2
:GHS07
Storage Temperature:Ambient
Frequently Bought Together