Order Entry
Czech Republic
ContactUsLinkComponent
2-Ethylphenylhydrazine hydrochloride 97%
Corrosive
Irritant
Health
Environment
2-Ethylphenylhydrazine hydrochloride 97%
Catalog # APOSOR0985-100G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
Health
Environment
2-Ethylphenylhydrazine hydrochloride 97%
Catalog # APOSOR0985-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :172.66 g/mol
Melting Pt:178 °C (dec.)
Storage Temperature:Ambient
EINECS:421-460-2
UN:3077
ADR:9+EHS,III
 :GHS05
 :GHS07
 :GHS08
 :GHS09
CAS Number:19398-06-2
 :58711-02-7
Formula:C₈H₁₃ClN₂
MDL Number:MFCD00071599