

3-Bromo-1-propanol 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:627-18-9
:
Formula:BrH₂CCH₂CH₂OH
MDL Number:MFCD00002942
:138.99 g/mol
Boiling Pt:70…72 °C/10mmHg
Density:1.59 g/cm³ (25 °C)
Flash Pt:93 °C
:GHS05
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together