
3-(3-Pyridyl)propionic acid 98%
Catalog # APOSOR0840-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:151.17 g/mol
Melting Pt:156…160 °C
:GHS07
Storage Temperature:Ambient
CAS Number:3724-19-4
Formula:C₈H₉NO₂
MDL Number:MFCD00274197