2,3-Dimethoxybenzaldehyde 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:86-51-1
:
Formula:C₉H₁₀O₃
MDL Number:MFCD00003309
:166.18 g/mol
Boiling Pt:137 °C
Melting Pt:49 °C
Flash Pt:113 °C
Storage Temperature:Ambient
Frequently Bought Together