Malonic acid
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 1,3-Propanedioic acid
- CAS Number:141-82-2
- Formula:C₃H₄O₄
- MDL Number:MFCD00002707
- Molecular Weight:104.06 g/mol
- Melting Pt:132…136 °C (dec.)
- Density:1.62 g/cm³ (25 °C)
- Flash Pt:>110 °C
- Storage Temperature:Ambient
- EINECS:205-503-0
3 Options Available Below
Product Details & Documents
Synonyms: 1,3-Propanedioic acid
- CAS Number:141-82-2
- Formula:C₃H₄O₄
- MDL Number:MFCD00002707
- Molecular Weight:104.06 g/mol
- Melting Pt:132…136 °C (dec.)
- Density:1.62 g/cm³ (25 °C)
- Flash Pt:>110 °C
- Storage Temperature:Ambient
- EINECS:205-503-0



