
4,6-Dimethyl-2-piperazin-1-ylpyrimidine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:22746-09-4
:
Formula:C₁₀H₁₆N₄
MDL Number:MFCD00885959
Boiling Pt:115…118 °C/1mmHg
:192.27 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together