Order Entry
Czech Republic
ContactUsLinkComponent
5-Methyl-1H-pyrazole-3-carboxylic acid
Irritant
5-Methyl-1H-pyrazole-3-carboxylic acid
Catalog # APOSOR0650-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-Methyl-1H-pyrazole-3-carboxylic acid
Catalog # APOSOR0650-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :126.11 g/mol
Melting Pt:236…240 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:402-61-9
 :MFCD00090754
Formula:C5H6N2O2
MDL Number:MFCD00462235